Learn More
Specac™ KBr Sealed Heatable Liquid Flow Cell VTCH Compatible
Brand: Specac™ GS20561
Code : Z3
Additional Details : Weight : 0.01000kg
Description
This cell is designed for use in the Specac variable temperature cell GS21525. This is a sealed heatable liquid flow cell with KBr windows and choice of pathlength from 0.025mm up to 1.0mm. This cell is fitted with 1/16in. Swagelok fittings for flow purposes.Specifications
Pathlength from 0.025 mm up to 1.0 mm |