missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KD Scientific Legato Syringe Pumps: Racks
For use with Legato Syringe Pumps
Brand: KD Scientific 788301
This item is not returnable.
View return policy
Description
- Accommodates up to ten 0.5 μL to 20 mL syringes or six 30 to 60 mL plastic or glass syringes
Certifications
CE Marked, UL Listed, CSA, WEE, CB scheme, EU RoHS
Specifications
0/1 V | |
CE, UL, CSA, WEE, CB scheme, EU RoHS | |
Legato syringe pump | |
4 |
0.5 μL to 140 mL | |
Small Syringe Multi-rack | |
±0.35% |