missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KDalert™ GAPDH Assay Kit

Product Code. 10535595
Click to view available options
Quantity:
375 Reactions
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10535595

Brand: Invitrogen™ AM1639

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

KDalert™ GAPDH Assay Kit

The Ambion™ KDalert™ GAPDH Assay Kit is for the reliable measure of GAPDH enzyme activity in cultured human, mouse, or rat cells in less than 30 minutes using a microplate fluorometer. The kit includes sufficient reagents for 375 reactions.

  • Assess GAPDH siRNA delivery in 1/3 the time for 1/3 the cost of real-time PCR
  • Analyze 1-96 samples simultaneously
  • Measure both GAPDH siRNA-induced knockdown AND transfection-induced toxicity
  • Compatible with a wide variety of cells and a broad range of culture conditions

    The KDalert GAPDH Assay Kit is an ideal positive control for transfection optimization experiments and also measures transfection induced cytoxicity. It is designed for use with Ambion™ Silencer™ GAPDH siRNA.

    Rapid, Time-Saving Procedure
    Use the assay to optimize siRNA transfection by transfecting individual cell samples with a GAPDH siRNA and a negative control siRNA. Two to three days after transfection, simply add the included cell lysis buffer to the cells, incubate for 20 minutes, add the diluted master mix of assay reagents, and read the increase in fluorescence four minutes later using a microplate or standard fluorometer. The assay procedure can be completed in about 30 minutes with minimal sample handling.

    One Assay for Two Readouts
    Because GAPDH is expressed at relatively constant levels, the assay can also be used to monitor transfection agent induced toxicity. For this analysis, GAPDH enzyme activity from negative control siRNA-transfected cells is compared to that of untreated cells. Reduced GAPDH activity in negative control-transfected cells compared to non-transfected cells is an indication that the transfection-induced cytotoxicity.

    Accessory Products:
    The KDalert™ Kit is designed for use with Silencer™ GAPDH siRNAs (SKUs #AM4605, AM4633, AM4634, AM4624, AM4632, or AM4631). Additional KDalert™ Lysis Buffer (SKU #AM8790G) is also available separately.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage KDalert Lysis Buffer can be stored at –20°C or 4°C. Solution A, Solution B, and Solution C should be stored at –20°C. GAPDH Enzyme should be stored at or below –70°C.
    Assay GAPDH Assay
    Detection Method Fluorescence
    Product Line Ambion, KDalert
    Product Type GAPDH Assay Kit
    Substrate Properties Chemical Substrate
    Sufficient For 375 Reactions
    For Use With (Application) Absorbance, Fluorescence Intensity
    Quantity 375 Reactions
    Shipping Condition Dry Ice
    Target GAPDH
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.