Learn More
DWK Life Sciences KECK™ Adapter KA, complete, from hose connection glass to connection plastic
KECK™ adapters prevent glass breakage on olives.
Brand: DWK Life Sciences 286344006
This item is not returnable.
View return policy
Description
KECK™ adapters prevent glass breakage on olives and protect fingers and hands from injuries. The clamping ring, O-ring and screw cap are permanently screwed to the glass olive. The hose part can be connected quickly, securely and tightly to the threaded cap with just 1/4 turn and can also be loosened again just as quickly.
- Quick connection and disconnection of the hose part to the threaded cap with only 1/4 turn
- permanent screw connection with the glass nozzle
- KECK™ adapters prevent glass breakage on nozzles and protect fingers and hands from injuries
Specifications
10 | |
Connection of a Hose Piece with a Threaded Cap | |
Connection of a hose piece with a threaded cap | |
GL 14, diameter 4.5 mm |