Learn More
DWK Life Sciences KECK™ Adapter KA, complete, from hose connection glass to connection plastic
KECK™ adapters prevent glass breakage on olives.
Brand:  DWK Life Sciences 286344109
 View more versions of this product
Product Code. 11771419
  • £79.20 / Pack of 10
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
KECK™ adapters prevent glass breakage on olives and protect fingers and hands from injuries. The clamping ring, O-ring and screw cap are permanently screwed to the glass olive. The hose part can be connected quickly, securely and tightly to the threaded cap with just 1/4 turn and can also be loosened again just as quickly.
  • Quick connection and disconnection of the hose part to the threaded cap with only 1/4 turn
  • permanent screw connection with the glass nozzle
  • KECK™ adapters prevent glass breakage on nozzles and protect fingers and hands from injuries
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10
Connection of a Hose Piece with a Threaded Cap
Connection of a hose piece with a threaded cap
GL 14, diameter 9.0 mm