Learn More
DWK Life Sciences KECK™ Clip Assortment, for conical joints, from metal
The KECK™ clamp is used to secure a conical ground joint connection.
Brand:  DWK Life Sciences 290330008
Product Code. 10019910
  • £75.40 / Each
Estimated Shipment: 18-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
KECK™ clamps are ground joint clamps used to secure standard ground joint connections in glass apparatus and are characterized by their easy handling and excellent resilience. The clamps are available in different materials (metal or polyoxymethylene).
  • For conical joints
  • Dimensionally stable with particularly good spring properties
  • Easy, safe and quick to set up
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Ea.
2 x KCM 14, 2 x KCM 19, 1 x KCM 29, 1 x KCM 24
Secure glass-to-glass joints
Metal
Secure Glass-to-Glass Joints