Learn More
DWK Life Sciences KECK™ Clip for Conical Joints, Ni, KCM
KECK™ conical ground joint clamps for high temperature applications.
Brand:  DWK Life Sciences 286337302
4.28 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : A0
Additional Details : Weight : 0.00540kg
 View more versions of this product
Product Code. 10579181
  • £5.50 / Each
Estimated Shipment: 23-08-2024
to see stock.
Add to basket
Description
Description
For applications at very high temperatures, the KECK™ nickel-plated spring steel ground joint clamps an important addition to KECK™ conical ground joint clips made of POM (polyoxymethylene). Compared to conventional wire clamps, they are easy and safe to attach and are inherently stable.
  • Easy and safe to put on and are dimensionally stable
  • Good dimensional stability compared to conventional wire clamps
Specifications
Specifications
100
For Use at Very High Temperatures
NS 14
Nickel
For use at very high temperatures; Securing conical ground joints