Learn More
DWK Life Sciences™ KECK™ Clip for Conical Joints, Ni, KCM
Shop All DWK Life Sciences Products
Click to view available options
Size:
NS 14
NS 19
NS 24
NS 29
Unit Size:
Each
Pack of 100
Description
For applications at very high temperatures, the KECK™ nickel-plated spring steel ground joint clamps an important addition to KECK™ conical ground joint clips made of POM (polyoxymethylene). Compared to conventional wire clamps, they are easy and safe to attach and are inherently stable.
- Easy and safe to put on and are dimensionally stable
- Good dimensional stability compared to conventional wire clamps
Specifications
Specifications
| Quantity | 100 |
| Material | Nickel |
| For Use With (Equipment) | For Use at Very High Temperatures |
| For Use With (Application) | For use at very high temperatures; Securing conical ground joints |
| Size | NS 29 |