missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences™ KECK™ Clip, for conical joints, from POM

Product Code. p-7189345 Shop All DWK Life Sciences Products
Click to view available options
Size:
NS 12
NS 14
NS 19
NS 24
NS 29
NS 34
NS 40
NS 45
Color:
Amber
Blue
Green
Orange
Red
Violet
Yellow
Unit Size:
Pack of 10

Product Code. 10643353

Brand: DWK Life Sciences™ 1090978

Only null left
Add to basket
Edge
Add to basket

The KECK™ clamp is used to secure a standard ground joint connection.

KECK™ clamps are ground joint clamps used to secure standard ground joint connections in glass apparatus and are characterized by their easy handling and excellent resilience. The clamps are available in different materials (metal or polyoxymethylene).

  • For conical joints
  • Dimensionally stable with particularly good spring properties
  • Easy, safe and quick to set up
  • Increased safety in the laboratory as the glass surface is not scratched
  • Marking of the individual sizes for better differentiation
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10
Material POM
For Use With (Equipment) Secure Glass-to-Glass Joints
For Use With (Application) Secure glass-to-glass joints
Size NS 14
Color Yellow
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.