missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences™ KECK™ Clip for Conical Joints, POM, KC
Shop All DWK Life Sciences Products
Click to view available options
Size:
NS 10
NS 12
NS 14
NS 19
NS 24
NS 29
NS 34
NS 40
NS 45
Color:
Amber
Blue
Green
Orange
Purple
Red
Yellow
Unit Size:
Each
Pack of 100
Description
KECK™ POM (polyoxymethylene) conical ground joint clamps are easy, safe and quick to attach and are characterized by their dimensional stability and good spring properties. In contrast to conventional wire clamps, the glass surface is not damaged when it is attached. This increases safety in the laboratory. The sizes can be easily distinguished from each other by the different colours.
- Excellent dimensional stability and good spring properties
- No damage to the glass surface during attachment unlike conventional wire clamps
- Easy, safe and quick to put on and take off
- Colour coding per size
Specifications
Specifications
| Quantity | 100 |
| Material | POM |
| For Use With (Equipment) | Securing of Conical Ground Joints |
| For Use With (Application) | Securing of conical ground joints |
| Size | NS 14 |
| Color | Yellow |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction