Learn More
DWK Life Sciences KECK™ Locking Ring for Adapter KA
Spare part for the KECK™ adapter KA from glass olive to threaded plastic nozzle.
Brand:  DWK Life Sciences 286339303
 View more versions of this product
Product Code. 15854688
  • £136.00 / Pack of 100
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The clamping ring is a component and therefore a spare part for the KECK™ Adapter KA from glass olive to plastic olive. It is available for threads GL 14 and RD 14.
  • The clamping ring is a component of the KECK™ Adapter KA and connects the glass olive with the plastic olive.
  • available for threads GL 14 and RD 14
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100
Sealing
Yellow
Sealing
GL 14