missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Keratinocyte-SFM, with L-glutamine
£748.00
Specifications
Cell Type | Keratinocyte Cells, Epithelial Cells |
---|---|
Classification | Serum-free |
Culture Type | Mammalian Cell Culture |
Form | Liquid |
Product Type | Serum Free Medium (SFM) |
Specifications
Keratinocyte Cells, Epithelial Cells | |
Mammalian Cell Culture | |
Serum Free Medium (SFM) | |
Sterile | |
Glutamine, Phenol Red |
Serum-free | |
Liquid | |
Serum-free | |
Sterile-filtered | |
Sustainable packaging |