missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kerosene, Reagent grade, Honeywell™ Riedel-de Haën™
Click to view available options
Quantity:
3.8 L
18.9 L
Packaging:
Glass Bottle
Stainless Steel Can
Description
- ≥95% for general laboratory use.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Colorless |
Density | 0.8g/mL at 25°C |
Boiling Point | 175°C to 325°C |
Flash Point | 70°C (158°F) |
Packaging | Stainless Steel Can |
Quantity | 18.9 L |
UN Number | UN1223 |
Vapor Density | 4.5 (vs air) |
Grade | Reagent |
Physical Form | Liquid |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'