missing translation for 'onlineSavingsMsg'
Learn More

Ketoketal, 97%, Thermo Scientific™

£16.78 - £61.41

Chemical Identifiers

CAS 56309-94-5
Molecular Formula C14H22O3
Molecular Weight (g/mol) 238.327
MDL Number MFCD00809704
InChI Key ZNWLFTSPNBLXGL-UHFFFAOYSA-N
Synonym ketoketal, 4-1,4-dioxaspiro 4.5 decan-8-yl cyclohexanone, 8-4-oxocyclohexyl-1,4-dioxaspiro 4.5 decane, 4-1,4-dioxaspiro 4.5 dec-8-yl cyclohexanone, 4-1,4-dioxaspiro 4.5 decan-8-yl cyclohexan-1-one, bicyclohexane-4,4'-dione monoethylene ketal, 1,1'-bicyclohexane-4,4'dione monoethylene ketal, dicyclohexane-4,4'dione monoethylene ketal, dicyclohexane-4,4'-dione monoethylene ketal, cyclohexanone, 4-1,4-dioxaspiro 4.5 dec-8-yl
PubChem CID 810913
IUPAC Name 4-(1,4-dioxaspiro[4.5]decan-8-yl)cyclohexan-1-one
SMILES C1CC(=O)CCC1C2CCC3(CC2)OCCO3
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15401208
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H55069.06
5 g
£16.78
5g
Please to purchase this item. Need a web account? Register with us today!
15411208
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H55069.14
25 g
£61.41
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ketoketal is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

56309-94-5
238.327
ZNWLFTSPNBLXGL-UHFFFAOYSA-N
810913
C1CC(=O)CCC1C2CCC3(CC2)OCCO3
C14H22O3
MFCD00809704
ketoketal, 4-1,4-dioxaspiro 4.5 decan-8-yl cyclohexanone, 8-4-oxocyclohexyl-1,4-dioxaspiro 4.5 decane, 4-1,4-dioxaspiro 4.5 dec-8-yl cyclohexanone, 4-1,4-dioxaspiro 4.5 decan-8-yl cyclohexan-1-one, bicyclohexane-4,4'-dione monoethylene ketal, 1,1'-bicyclohexane-4,4'dione monoethylene ketal, dicyclohexane-4,4'dione monoethylene ketal, dicyclohexane-4,4'-dione monoethylene ketal, cyclohexanone, 4-1,4-dioxaspiro 4.5 dec-8-yl
4-(1,4-dioxaspiro[4.5]decan-8-yl)cyclohexan-1-one

Specifications

56309-94-5
MFCD00809704
Insoluble in water.
C1CC(=O)CCC1C2CCC3(CC2)OCCO3
238.327
238.33
Ketoketal
C14H22O3
ketoketal, 4-1,4-dioxaspiro 4.5 decan-8-yl cyclohexanone, 8-4-oxocyclohexyl-1,4-dioxaspiro 4.5 decane, 4-1,4-dioxaspiro 4.5 dec-8-yl cyclohexanone, 4-1,4-dioxaspiro 4.5 decan-8-yl cyclohexan-1-one, bicyclohexane-4,4'-dione monoethylene ketal, 1,1'-bicyclohexane-4,4'dione monoethylene ketal, dicyclohexane-4,4'dione monoethylene ketal, dicyclohexane-4,4'-dione monoethylene ketal, cyclohexanone, 4-1,4-dioxaspiro 4.5 dec-8-yl
ZNWLFTSPNBLXGL-UHFFFAOYSA-N
4-(1,4-dioxaspiro[4.5]decan-8-yl)cyclohexan-1-one
810913
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H317
May cause an allergic skin reaction.

P261-P272-P280g-P302+P352-P333+P313-P363-P501c

H317

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications