missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ KingFisher™ Plastics for 24 deep-well format

Product Code. 12663116 Shop All Thermo Scientific Products
missing translation for 'orderingAttributeHoverText'
Product Type:
24 Combi tip comb
24 DW plate, barcoded, sterile
24 DW tip comb and plate
24 DW tip comb, plate, barcode
24 DW tip comb, plate, sterile
24 Deep Well plate, barcoded
24 Deep-well Plate
24 Deep-well Plate, Sterile
6-tip comb and 24 DW plate
For Use With (Equipment):
KF Duo Prime, Flex, Presto
KingFisher Apex
KingFisher Duo Prime
KingFisher Flex and Presto
missing translation for 'unitSize'
Pack of 48
Pack of 50
This item is not returnable. View return policy

Product Code. 12663116

missing translation for 'mfr': Thermo Scientific™ 95040480

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Flex, Duo Prime, and Presto instruments.

Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.

Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Flex, KingFisher Duo Prime, and KingFisher Presto.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

TRUSTED_SUSTAINABILITY

Specifications

Product Type 24 Deep-well Plate, Sterile
Format 24 Deep-well Plate
Capacity 24 Samples
For Use With (Equipment) KF Duo Prime, Flex, Presto

For research use only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.