Learn More
Thermo Scientific™ KingFisher Apex Accessories
KingFisher Apex is a flexible system for different types of sample processing needs which offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein and cell purification from virtually any source.
Brand:  Thermo Scientific™ 24075930
 View more versions of this product
Product Code. 16512442
  • £608.00 / Each
Explore more special offers
Description
Description
KingFisher Apex is a flexible system for different types of sample processing needs which offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein and cell purification from virtually any source. Using revolutionary magnetic particle separation technology, this system provides excellent reproducibility and quality
  • User-friendly high throughput sample purification system for automating DNA/RNA, protein and cell purification
  • Provides versatile and high-speed purification with reproducible results
  • Easy to use interface with touch screen empowering the user to control all details of the application and purification process
  • BindIx PC Software for added convenience
  • Extensive selection of validated protocols for different type of applications
  • Cloud library for quick protocol retrieval
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Heating Block
96 Samples
Reusable
1 Pc.
96 Deep-well Heating Block
KF Apex 96 DW heating block
KingFisher Apex