Learn More
KG-501, MedChemExpress
MedChemExpress KG-501 is a CREB inhibitor, with an IC50 of 6.89 μM.
Brand:  Medchem Express HY-103299
Encompass_Preferred
 View more versions of this product
Product Code. 16460197
  • £200.86 / 50mg
Explore more special offers
Description
Description
KG-501 is a CREB inhibitor, with an IC50 of 6.89 μM.
Specifications
Specifications
KG-501
Light Yellow
C17H13ClNO5P
Naphthol AS-E phosphate
O=C(C1=C(OP(O)(O)=O)C=C2C=CC=CC2=C1)NC3=CC=C(Cl)C=C3
377.72
Research
COVID-19-immunoregulation
18228-17-6
Research
50 mg
DMSO : 6 mg/mL (15.88 mM; Need ultrasonic and warming)
377.72
98.08%
Solid