Learn More
Lonza™ KGM™-2 Keratinocyte Growth Medium-2 SingleQuots™ Supplements and Growth Factors
Supplements and growth factors kit for use with keratinocyte basal medium.
Brand: Lonza™ CC-4152
Additional Details : Weight : 0.00050kg Transport : UN number : 1170 Chem class : 3 Pack group : II
Description
- Separately sold SingleQuot™ kit for supplementing basal medium.
- Part of the full KGM™-2 Keratinocyte Growth Medium-2 BulletKit™, which consists of basal medium (CC-3103) and supplements packaged together.
- Includes the following:
- BPE Low Protein - non specific
- HEGF
- Recombinant Human Insulin (0.5%)
- Hydrocortisone Solution
- Transferrin
- Epinephrine
- GA-1000
Specifications
BPE Low Protein - non specific, HEGF, Recombinant Human Insulin 0.5%, Hydrocortisone Solution, Transferrin, Epinephrine, GA-1000 | |
Keratinocyte Cells | |
Supplements and growth factors |