Learn More
Lonza™ KGM™ Gold Keratinocyte Growth Medium BulletKit™
A new robust and versatile serum-free medium formulation for high and low density NHEK-Normal Human Epidermal Keratinocyte cultures.
Brand: Lonza™ 192060
Code : NEW
Additional Details : Weight : 0.58000kg Transport : UN number : 1170 Chem class : 3 Pack group : II
Description
- Designed to support both clonal growth and high density keratinocyte proliferation.
- Supports superior growth and morphology of keratinocytes.
- Quality control requiring that keratinocytes pass strict morphological criteria specifying the minimal presence of enlarged and vacuolated cells, enabling Lonza keratinocytes to be used by the researcher for ≥15 population doublings.
- 1 x KBM™Gold™ Basal Medium (00192151), 500 mL
- 1 x KGM™Gold™SingleQuots™Supplement pack (00192152) containing:
1 x Natural Cap Vial with Hydrocortisone, 0.50 mL
1 x Natural Cap Vial with Transferrin, 0.50 mL
1 x Amber Vial with Epinephrine, 0.25 mL
1 x Red Cap Vial with GA-1000, 0.50 mL
1 x Orange Cap Vial with BPE, 2.00 mL
1 x Green Cap Vial with hEGF, 0.50 mL
1 x Lilac Cap Vial with Insulin, 0.50 mL
Specifications
KBM Gold Basal Medium, KGM Gold SingleQuots Supplement Pack | |
Keratinocyte Cells | |
500 mL Bottle, 0.5 mL to 2.0 mL Vials |
Safety and Handling
- KGM Gold SingleQuots
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash skin thoroughly after handling.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P363-Wash contaminated clothing before reuse.
- P370+P378-In case of fire: Use water spray, alcohol-resistant foam, drychemical or carbondioxide to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol, Gentamicinsulfate