Learn More
Unity Lab Services™ KI Test for Biosafety Cabinet (≥ 1.8 m2)
Product Code. 17899602
missing translation for 'orderingAttributeHoverText'
Duration:
1 Day
missing translation for 'unitSize'
Each
Strongly recommended (and sometimes required) in addition to basic certification (part no BSCCERT)
Product Code. 17899602
missing translation for 'mfr': Unity Lab Services™ BSCKI18L
Strongly recommended (and sometimes required) in addition to basic certification (part no BSCCERT)
European KI test for biosafety cabinets 1.8 mt or wider; Equipment is required to pass certification prior to this service. Required for UK. This is add-on to STARTBSC[PF]
  • Equipment is required to pass EN12469 certification prior to this test
  • Required for UK
  • This is add-on to STARTBSC[PF] for new equipment, and an add on to BSCCERT for existing equipment
  • Includes travel and labor unless extensive travel is needed
  • All additional works, as i.e. waiting time or correctional works, will be invoiced additionally according to our recent valid service pricelist
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Biosafety Cabinets
Certifications/Compliance BS EN 12469 (Europe) or NSF 49 (US)
Duration 1 Day
Product Type European KI Test
Product Line Thermo Scientific
Service Offering Performance Test