missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human KIM-1 (TIM-1/HAVCR1) ProcartaPlex™ Simplex Kit

The Human KIM-1 (TIM-1/HAVCR1) ProcartaPlex Simplex Kit measures KIM-1 (TIM-1/HAVCR1) protein and is designed to be combinable with other Simplex kits so that users can create their own multiplex panel that utilizes Luminex xMAP technology for protein detection/quantitation.

Brand:  Invitrogen™ EPX010-12445-901

Additional Details : Weight : 0.20000kg

Product Code. 17857382

  • £221.00 / Pack of 96
Estimated Shipment: 26-07-2024
to see stock.

Description

Description

The Human KIM-1 (TIM-1/HAVCR1) ProcartaPlex Simplex Kit measures KIM-1 (TIM-1/HAVCR1) protein and is designed to be combinable with other Simplex kits so that users can create their own multiplex panel that utilizes Luminex xMAP technology for protein detection/quantitation. When combining multiple Simplex kits (i.e., when not using a pre-configured Multiplex Panel), only one buffer kit (sold separately) is needed for each assay plate regardless of plex size.

About ProcartaPlex assays for the Luminex platform
ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant, and just four hours to obtain analyzed results.

Features include:
More results per sample—measure multiple protein targets simultaneously in a single 25–50 μL sample
Well-established Luminex technology—highly referenced multiplexing platform for protein detection and quantitation

ProcartaPlex assays utilize Luminex xMAP (multianalyte profiling) technology for the simultaneous detection and quantitation of up to 65 protein targets in a single 25–50 μL sample—from plasma, serum, cell culture supernatants, and other bodily fluids.

The Luminex beads in the ProcartaPlex assay are internally dyed with precise proportions of red and infrared fluorophores to create spectrally unique signatures that can be identified by the Luminex xMAP detection systems (e.g., Luminex 200, FLEXMAP 3D, and MAGPIX). Similar to a sandwich ELISA, the ProcartaPlex assay uses matched antibody pairs to identify the protein of interest. In a multiplexed assay, each spectrally unique bead is labeled with antibodies specific for a single target protein, and bound proteins are identified with biotinylated antibodies and streptavidin–R-phycoerythrin (RPE). The conjugation of protein-specific antibodies to a distinct bead allows for analysis of multiple targets in a single well.

The most significant difference between a ProcartaPlex assay and ELISA is that the capture antibody in the ProcartaPlex assay is conjugated to a bead and not adsorbed to the microplate well, so the ProcartaPlex assay reagents are free-floating in the solution. For detection, the Luminex 200 instrument, for example, contains two lasers, one to distinguish the spectral signature of each bead and the second to quantify the amount of RPE fluorescence, which is proportional to the amount of protein present in the sample. ProcartaPlex multiplex assays can profile more target proteins using significantly less sample in the same time that it takes to perform a traditional sandwich ELISA.

ProcartaPlex multiplex panels are available in multiple formats across six species (human, mouse, rat, nonhuman primate, porcine, and canine).

Specifications

Specifications

Simplex Kit
Human
Magnetic
As determined for Lot 1: 164.6 pg/mL
Simplex Kit (combinable)
HAVCR1
26762
As determined for Lot 1: 3.9%
ProcartaPlex™
Urine, Serum, Plasma: 25 μL
CCS: 50 μL
1 vial Capture Beads (50x)
1 vial biotinylated Detection Antibody (50x)
2 vials Standard Mix (lyophilized)

Store at 2-8°C.
Immunology, Toxicology
96 Tests
Combinable
KIM-1 (TIM-1/HAVCR1) [53]
See product documentation
Luminex™ Instruments
HAVCR1, HAVCR, HAVCR-1, KIM-1, KIM1, TIM, TIM-1, TIM1, TIMD-1, TIMD1, CD365, hepatitis A virus cellular receptor 1
Yes (384-well Mix and Match panel)
As determined for Lot 1: 4.9%
Urine, Serum, Plasma, Cell Culture Supernatants (CCS)
Q96D42
Wet Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers