missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kim-Kap™ Disposable Closures

Available in natural (clear) or choice of four colors

Brand:  DWK Life Sciences 73660-25

Product Code. 10054454

  • £140.00 / Pack of 500
Estimated Shipment: 14-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Available in natural (clear) or choice of four colors

  • For plain-end culture tubes
  • Keep tube contents sterile without need for cotton plugging
  • Polypropylene
  • Autoclavable

Caps for 38mm O.D. are designed for baffled culture flasks.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Closure
Polypropylene
25 mm O.D. Plain-end Culture Tubes
Autoclavable
Natural
Single-use
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers