missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimberly-Clark Professional™ KleenGuard™ G60 Medium Duty Polyurethane-Coated Cut-Resistant Gloves
KleenGuard™ G60 Medium Duty Polyurethane-Coated Cut-Resistant Gloves are constructed of ultra-high molecular weight polyethylene fibers to enhance breathability, and provide ANSI Cut Level 3 protection.
Specifications
ANSI Abrasion Rating | ANSI 4 |
---|---|
Coating Material | Polyurethane |
Color | Gray |
Cut Resistance Rating | ANSI A2 |
Hand | Specific |
Description
- Constructed of ultra-high molecular weight polyethylene (UHMWPE) fibers to provide breathability and protection
- Polyurethane coated palm and fingers enhance dexterity and grip capabilities
- Separate right and left gloves in a pair provide a snug fit
Specifications
ANSI 4 | |
Gray | |
Specific | |
HPPE | |
EN 388 | |
Knit | |
13 |
Polyurethane | |
ANSI A2 | |
Yes | |
Cut-resistant Gloves | |
Assembly, plastics, light raw materials | |
KleenGuard™ G60 |