missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark Professional™ KleenGuard™ G10 Arctic Blue Nitrile Gloves

KleenGuard™ G10 gloves provide durability and strength for light-duty tasks. Ideal for industrial, manufacturing and food processing applications.

Specifications

Certifications/Compliance FDA 21 CFR 170-199, CFIA approved
Applications Industrial
AQL 4
Color Blue
Disposable Single-use
View More Specs

Description

Description

  • Powder free, nitrile
  • Textured fingertips provide enhanced grip and dexterity
  • Ambidextrous
  • Beaded cuff
  • Puncture-resistant formulation delivers barrier protection against chemical splash, dirt, grease and oil
  • Ideal for cleaning and maintenance, manufacturing, food processing, electronic assembly and painting applications
Specifications

Specifications

FDA 21 CFR 170-199, CFIA approved
4
Single-use
Yes
Powder Free
0.05 mm
24 cm
Industrial
Blue
Partially Textured
Nitrile
2 mil
9.5 in.
Videos
SDS
Documents

Documents

Product Certifications