missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Black Phenolic Screw-Thread Closures

Highly resistant to temperature and steam during autoclaving

£134.00 - £425.00

Specifications

Color Black
Material Phenolic Resin
Autoclavable Autoclavable
Product Type Screw Cap
For Use With (Equipment) Bottle
View More Specs

Products 2
Product Code Brand Neck Finish Price Quantity & Availability  
Product Code Brand Neck Finish Price Quantity & Availability  
10455342
View Documents
DWK Life Sciences
73802-15415
15-415
£425.00
Pack of 500
In Stock
to see stock.
Add to basket
 
11894005
View Documents
DWK Life Sciences
73802-13415
13-415
£134.00
Pack of 500
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
 
Description

Description

  • Cemented-in PTFE-faced 14B white rubber liner
  • Three screw-thread finishes

Specifications

Specifications

Black
Autoclavable
Bottle
Phenolic Resin
Screw Cap
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences Kimble™ Black Phenolic Screw-Thread Closures

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.