Learn More
DWK Life Sciences Kimble™ BioMasherII™ Closed System Micro Tissue Homogenizer
Ensure efficient grinding of samples by minimizing potential exposure to hazardous materials
Brand: DWK Life Sciences 9749625002
Code : NEW
Additional Details : Weight : 0.18800kg
Description
Ensure efficient grinding of samples by minimizing potential exposure to hazardous materials
- Clear polypropylene tube
- Polyacetal pestle with molded-in shaft guard
- Abrasive surface (pestle tip and inner tube area)
- Available in Sterile and non-sterile versions
- Capacity: 1.5mL
For animal tissues and organs, plant materials, insects, DNA, RNA, proteins, yeasts and enzymes
Specifications
Polypropylene tube, Polyacetal pestle | |
Clear | |
1.5 mL | |
Sterile | |
For animal tissues and organs, plant materials, insects, DNA, RNA, Proteins, yeasts and Enzymes |
0.05 oz., 1.5mL | |
Single pack | |
0.05 oz. | |
Yes |