Learn More
DWK Life Sciences Kimble™ Economy Columns Sample Diffusion Disc
Product Code. 13476349
Click to view available options
Diameter (Metric) Inner:
1 cm
Unit Size:
Each
Product Code. 13476349
Brand: DWK Life Sciences 4204041000
Only null left
Add to basket
Add to basket
Flex-Column™ sample diffusion discs protect the gel bed from being disrupted during sample application whenever a flow adapter is not used.
  • A transparent acrylic ring is sealed to a 20um porosity HDPE disc which protects the top of the gel bed when the sample is manually applied
  • A crossbar on the top of the acrylic ring permits easy removal of the sample diffusion disc from the column.
TRUSTED_SUSTAINABILITY
Specifications
Type Sample Diffusion Disc
Diameter (Metric) Inner 1 cm
For Use With Kimble-Chase Economy Columns