Learn More
DWK Life Sciences Kimble™ Economy Columns Sample Diffusion Disc
Click to view available options
Diameter (Metric) Inner:
1 cm
Unit Size:
Each
Description
Flex-Column™ sample diffusion discs protect the gel bed from being disrupted during sample application whenever a flow adapter is not used.
- A transparent acrylic ring is sealed to a 20um porosity HDPE disc which protects the top of the gel bed when the sample is manually applied
- A crossbar on the top of the acrylic ring permits easy removal of the sample diffusion disc from the column.
Specifications
Specifications
| Type | Sample Diffusion Disc |
| Diameter (Metric) Inner | 1 cm |
| For Use With | Kimble-Chase Economy Columns |