Learn More
DWK Life Sciences Kimble™ KIMAX™ Pycnometer Bottle
Designed for measuring and comparing the densities or specific gravities of liquids and solids. This non-mercury spirit-filled pycnometer is supplied with Standard Taper 10/18 thermometer with a range of 14 to 38°C in 0.2° increments.
Specifications
Product Type Pycnometer Bottle
View More Specs
Description
Description
  • Thermometer is retested for accuracy within ±0.2 degrees C
  • Standard Taper 5/12 side tube cap has a small vent near the top
  • Body is conical for maximum stability
  • Capacity is within 10 percent of nominal shown on the flask
  • Each flask body is numbered for ready identification
Specifications
Specifications
Pycnometer Bottle
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
DWK Life Sciences Kimble™ KIMAX™ Pycnometer Bottle