Learn More
DWK Life Sciences Kimble™ Kontes™ Chromaflex™ Columns
Ideal for medium-pressure chromatography with aqueous mobile phases
Brand:  DWK Life Sciences 420830-1510
 View more versions of this product
Product Code. 15963470
  • £340.00 / Each
Estimated Shipment: 11-03-2025
to see stock.
Explore more special offers
Add to basket
Includes: Glass barrel, Two PTFE end fittings with 20μm porosity polyethylene bed supports, 5 ft. (1.5m) of PTFE tubing (1/16 in. O.D. tubing with 1.0cm I.D. columns, 1/8 in. O.D. tubing with 2.5 and 4.8cm I.D. columns), Two flangeless tubing nuts, Two flangeless ferrules
Description
Description
Ideal for medium-pressure chromatography with aqueous mobile phases
  • Threaded
  • Feature PTFE-shielded O-rings and end fittings
  • PTFE layer is curled over the O-ring to form a solvent- and leak-resistant seal
  • Solvents contact only PTFE and borosilicate glass surfaces
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Medium pressure chromatography with aqueous mobile phases
25 mm
75.41 psi
74 mL
Borosilicate Glass Type I Class A
1/4-28 Flangeless fitting
Ethylene Oxide, 2N NaOH or 100% Ethanol
0 to 50°C
Chromatography Column
20 μm
150 mm
HDPE
Two PTFE end fittings with 20um porosity Polyethylene bed supports, 5 ft. PTFE tubing, 2 flangeless tubing nuts and 2 flangeless ferrules