Learn More
DWK Life Sciences Kimble™ Kontes™ Chromatography Column Accessories
For chromatography columns
£18.00
Specifications
For Use With (Equipment) For chromatography columns
Thread Size 13-425
View More Specs
Products 1
Product Code Brand Product Type Price Quantity & Availability  
Product Code Brand Product Type Price Quantity & Availability  
17790878
View Documents
DWK Life Sciences
747300-0050
Drying Tube/Chromatography Column
£18.00
Each
Estimated Shipment: 04-09-2025
to see stock.
Add to basket
Description
Description
  • 747000 Series Microflex Kits
  • Features versatile threaded chaste connections
  • Can be assembled rightside up or upside down, providing great versatility
  • GPI thread size: 13 to 425
Specifications
Specifications
For chromatography columns
13-425
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
DWK Life Sciences Kimble™ Kontes™ Chromatography Column Accessories