Learn More
DWK Life Sciences Kimble™ Kontes™ Chromatography Cylindrical TLC Developing Tanks
Cylindrical glass tanks for the development of 1 x 3 in. to 10 x 20cm plates.
Brand:  DWK Life Sciences 416170-1020
 View more versions of this product
Product Code. 13055803
  • £228.00 / Each
Estimated Shipment: 10-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • These tanks provide maximum vapor phase saturation with a minimum amount of solvent
  • Polyethylene or glass cap included
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Glass
Cylindrical TLC Developing Tank
10 x 20 cm Plate Size
Cylindrical
Glass
Provides Maximum Vapor Phase Saturation with a Minimum Amount of Solvent
10 H x 5 cm I.D. Plate