Learn More
DWK Life Sciences Kimble™ Kontes™ Extraction Manifold Replacement Components
For extraction manifold.
Brand: DWK Life Sciences 971100-0047
This item is not returnable.
View return policy
Description
- For use with Kontes SPE Manifold Sets (971000-1047, -3047, or -6047)
- Kontes SPE Glassware and Manifolds are specially designed as support systems for 47mm and 90mm dia. solid phase extraction disks to give you the highest possible recoveries of extracted analytes from your drinking water, surface water and waste water samples
Specifications
Solid Phase Extraction Manifold | |
47 mm Funnel, support base, support screen, PTFE gaskets and clamp | |
47 mm |
SPE Glassware Set (47 mm Funnel, support base, support screen, PTFE gaskets and clamp) | |
Glass |