missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Microfiltration Assembly Hardware
Use Kimble™ Microfiltration Assembly Hardware when filtering large volumes of viscous or proteinaceous solutions to give the maximum flow rate.
Description
- Microfiltrational assembly 1000mL
- Funnel, 4000mL
- Flask, borosilicate glass, 47mm
- Glass support and ground joint 90mm
- Anodized aluminum clamp and filtration flask
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title