Learn More
DWK Life Sciences Kimble™ Kontes™ Microcaps™ TLC Spotting Capillaries
For spotting preadsorbent TLC plates
£60.65 - £198.00
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
13005813
|
DWK Life Sciences
764500-0005 |
5 μL |
£156.00
Pack of 100 |
Estimated Shipment: 17-09-2024
Log in to see stock. |
|||||
13025813
|
DWK Life Sciences
764500-0020 |
20 μL |
£60.65
Pack of 100 |
Estimated Shipment: 17-09-2024
Log in to see stock. |
|||||
13065813
|
DWK Life Sciences
764500-0100 |
100 μL |
£60.65
Pack of 100 |
Estimated Shipment: 17-09-2024
Log in to see stock. |
|||||
13075813
|
DWK Life Sciences
764520-0000 |
0.5, 1, 2, 5, 10 μL |
£198.00
Pack of 500 |
Estimated Shipment: 17-09-2024
Log in to see stock. |
|||||
Description
Precision-bore glass capillary tubes, cut to predetermined lengths, so that each capillary tube will hold a known volume of fluid when filled.
- Capillary action draws fluid into tube
- Dispense fluid by squeezing bulb
- No unusual skills or training required
- Immediate, professional results
- Supplied in packages containing one bulb assembly and one dispenser vial with 100 micropipets
- Volumetric tolerance: ±1%
Specifications
Borosilicate Glass |