missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ Microscale Organic Chemistry LABSETS™ and Kits Components

Components for microscale organic chemistry LABSETS

Specifications

For Use With (Application) Microscale organic chemistry
View More Specs

Description

Description

  • Microflex Kits feature special chaste connectors and specially tooled glassware components, providing great versatility
  • They fit easily into lab bench drawers

Specifications

Specifications

Microscale organic chemistry
Videos
SDS
Documents

Documents

Product Certifications