missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ MIDI-VAP 4000 Component Replacement Parts
Component parts for both ammonia and phenol distillation system
£86.35
Product Code | Brand | Type | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | For Use With (Equipment) | Price | Quantity & Availability | ||||
16577279
|
DWK Life Sciences
479455-0050 |
Reaction/Absorption Tube | MIDI-VAP™ 4000 cyanide distillation system |
£86.35
Each |
Estimated Shipment: 30-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Component parts for both ammonia and phenol distillation system
- For use with MIDI-VAP 4000 Distillation Apparatus
Specifications
Distillation systems |