missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Packing Reservoir for CHROMAFLEX™ Columns
CHROMAFLEX™ packing reservoirs are ideal for packing gel slurries into columns
£249.00 - £495.00
Specifications
Packaging | End fitting has 1/4-28 threads for easy connection to pumps |
---|---|
Type | Packing Reservoir |
Product Code | Brand | Diameter (Metric) | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Quantity | Price | Quantity & Availability | ||||
15310168
|
DWK Life Sciences
420837-0000 |
1 cm | 150 mL |
£249.00
Each |
Estimated Shipment: 30-10-2024
Log in to see stock. |
||||
15350168
|
DWK Life Sciences
420837-0020 |
2.5 cm | 500 mL |
£495.00
Each |
Estimated Shipment: 19-11-2024
Log in to see stock. |
||||
Description
- Fits Standard Column
- Each packing reservoir is designed for easy mounting on the CHROMAFLEX™ columns
- The capacities are matched to take the amount of gel slurry required to pack the longest column
- The packing reservoirs may also be used as buffer reservoirs for gravity flow chromatography
- 1.0cmID column has nominal volume of 150mL
- 2.5cmID column has nominal volume of 500mL
- Temperature Range 0 to 50°C
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
End fitting has 1/4-28 threads for easy connection to pumps | |
Packing Reservoir |