missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ Pellet Pestle™

Use to resuspend protein and DNA pellets or grind soft tissue in microcentrifuge tubes

£203.00 - £363.00

Specifications

For Use With (Application) Resuspend Protein and DNA pellets or grind soft tissue in microcentrifuge tubes
View More Specs

Products 3
Product Code Brand Capacity (Metric) Material Includes Dnase Free Price Quantity & Availability  
Product Code Brand Capacity (Metric) Material Includes Dnase Free Price Quantity & Availability  
11872913
View Documents
DWK Life Sciences
749520-0000
1.5 mL Polypropylene 100 Microtubes No N/A N/A N/A
10794783
View Documents
DWK Life Sciences
749520-0590
0.5 mL Polypropylene 100 Microtubes RNase, DNase and Pyrogen-free
£203.00
Pack of 100
Estimated Shipment: 08-08-2025
to see stock.
Add to basket
 
11815125
View Documents Promotion Details
DWK Life Sciences
749521-1590
1.5 mL Polypropylene w/o Microtubes RNase, DNase and Pyrogen-free
£225.68
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

Use to resuspend protein and DNA pellets or grind soft tissue in microcentrifuge tubes

Disposable Pellet Pestle

  • Choose autoclavable polypropylene or RNase-, DNase-, pyrogen-free material (manufactured under M5.5 [Class 10,000] cleanroom conditions)
  • Available with or without 100 microtubes
  • Microtubes have the same properties and come individually wrapped to ensure purity
  • Length of pestle with tip, 7cm (2.75 in.)
Reusable Pellet Pestle
  • Autoclavable CTFE polymeric head with a stainless-steel shaft
  • Pestle length with tip, 6cm (2.5 in.) for 0.5mL size; 14cm (5.59 in.) for 1.5mL size

Specifications

Specifications

Resuspend Protein and DNA pellets or grind soft tissue in microcentrifuge tubes
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences Kimble™ Kontes™ Pellet Pestle™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.