missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Duall™ All-Glass Tissue Grinder Accessory, Grinding Tube

Improves grinding efficiency

Specifications

Product Type Tissue Grinder Tube
For Use With (Application) Improves grinding efficiency
For Use With (Equipment) Duall™ all Glass tissue grinders
View More Specs

Products 2
Product Code Brand Includes Size Price Quantity & Availability  
Product Code Brand Includes Size Price Quantity & Availability  
10009701
View Documents
DWK Life Sciences
885452-0022
Without plastic safety coating 22 N/A
N/A
12770368
View Documents
DWK Life Sciences
885452-0020
Without plastic safety coating 20 N/A
N/A
Description

Description

Improves grinding efficiency

  • Tubes sizes from 21 to 25
  • Available with or without coating
Specifications

Specifications

Tissue Grinder Tube
Improves grinding efficiency
Duall™ all Glass tissue grinders
Videos
SDS
Documents

Documents

Product Certifications