missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Disposable Polystyrene Serological Pipets in Canister

For mass testing and serological work.

Brand:  DWK Life Sciences 56700-10110

159.10 GBP valid until 2025-12-31
Use promo code "24326" to get your promotional price.


 View more versions of this product

Product Code. 16438955

  • £170.00 / Pack of 500
Estimated Shipment: 16-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Calibrated to deliver to the tip
  • Features two rings near the top
  • Irradiation sterilized
  • Printed with chemically resistant organic black ink

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Yes
Sterile
Yes
3 mL
±0.2 mL
0.33 oz.
Serological Pipet
Plastic
For mass testing and serological work
0.1 mL
500/Cs.
Canister
10 mL
Single-use
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.