Learn More
DWK Life Sciences KIMBLE™ EPA Water Analysis Vial, Without Closure
These clear and amber screw thread EPA vials for the collection and storage of water samples comply with the guidelines for establishing test procedures for the analysis of pollutants.
£482.00
Specifications
Material | Borosilicate Glass |
---|---|
Closure Type | Screw Cap |
Certifications/Compliance | EPA 601, 602, 603, 604 |
Description
- Clear EPA vials are Converted from Type I, 33 expansion borosilicate glass tubing and amber EPA vials are converted from Type I, 51 expansion borosilicate glass tubing
- While vials are not pre-cleaned, they are packaged in a plastic shrink wrap module, lehr-clean, as they come from the end of the production lehr and then are placed into a corrugated shelf-pack
- Open-top closures and PTFE-faced silicone septa or solid-top PFTE-faced white rubber-lined closures are available separately
Compliance
Vials comply with guidelines for establishing test procedures for analyzing pollutants in water (Ref. U.S. EPA Methods 601, 602, 603, and 604).
Specifications
Borosilicate Glass | |
Screw Cap | |
EPA 601, 602, 603, 604 |