missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ KIMAX™ 5-Place NMR Tube Washers

These complete five-place washers use flexible PTFE tubing and a vacuum source (not included) to direct a power stream of wash solvent to thoroughly clean the insides of 7 and 8 in. NMR tubes.

Brand:  DWK Life Sciences 897033-0005

 View more versions of this product

Product Code. 14820422

  • £692.00 / Each
Estimated Shipment: 17-09-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

These complete five-place washers use flexible PTFE tubing and a vacuum source (not included) to direct a power stream of wash solvent to thoroughly clean the insides of 7 and 8 in. NMR tubes.

  • Two sizes available for 3 and 5mm diameter tubes
  • Both sizes come with a 1L reservoir bottle
  • All wetted parts are constructed of chemical-resistant PTFE or borosilicate glass
  • An O-ring slips over each tube to form a vacuum-tight seal before tubes are positioned for cleaning
  • With a simple twist, the wash valve can be configured to wash and then dry
  • Cycles can be repeated, or cleaned tubes can be removed immediately for use
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PTFE or Borosilicate Glass
NMR Tube Washer
5 mm dia. Tubes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.