missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ KIMAX™ Flasks with standard taper Joints

Short Necks

Brand:  DWK Life Sciences 25285-1000

Additional Details : Weight : 4.00000kg

 View more versions of this product

Product Code. 17862811

  • £477.00 / Pack of 12
Estimated Shipment: 10-02-2025
to see stock.
Add to basket

Description

Description

Round bottoms

Compliance

Designed from ASTM™ E 1403, Type II, Class 3 requirements

Specifications

Specifications

5.1 in.
Borosilicate Glass
19 cm
24/40
130 mm
1000 mL
7.4 in.
Short
Round
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers