Learn More
DWK Life Sciences Kimble™ Kontes™ Closed System Sterile Tissue Grinder (Molded Tip)
Minimizes the risk of personal contact with samples
£153.00
Specifications
Includes | Fully assembled, 30% Glass filled Polypropylene pestle, Polypropylene conical sample tube with solid HDPE cap and an adhesive label |
---|---|
Material | Polypropylene |
Sterility | Sterile |
Description
Minimizes the risk of personal contact with samples
- Tissues are ground within a sealed container
- Narrow pestles eliminate the potential of sample overflow
- Molded-in abrasive surface on the pestle tip leaves no sediment to obstruct sample examination
- Gamma sterilized in easy-open HDPE packs
- CS2 tissue grinders have a vitrified tip for more aggressive grinding of difficult samples
- Polypropylene pestles and conical sample tubes with solid HDPE caps
Specifications
Fully assembled, 30% Glass filled Polypropylene pestle, Polypropylene conical sample tube with solid HDPE cap and an adhesive label | |
Polypropylene | |
Sterile |