missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Reagent Sprayers with Screw Thread Ground Joint
Provide uniform spray mist of visualization reagents for TLC and paper chromatography; use with all standard TLC reagents and most corrosive liquids
Brand: DWK Life Sciences 422530-0050
This item is not returnable.
View return policy
Description
Provide uniform spray mist of visualization reagents for TLC and paper chromatography; use with all standard TLC reagents and most corrosive liquids
- Liquid touches only nonreactive borosilicate glass atomizing sprayer head and flask
- Sprayer head-to-flask connection consists of screw-thread standard taper ground joint, connecting cap, and loosening ring
- Operate sprayer at 3 to 5psi (0.021 to 0.035MPa)
- Adjust spray pattern by varying the position of thumb on vent hole
- Autoclavable (after cap, loosening ring, and O-ring are removed)

Specifications
Autoclavable | |
50 mL | |
Glass | |
TLC Reagent Sprayer | |
Standard TLC Reagents and Most Corrosive Liquids | |
19/22 | |
17.4 cm | |
5 cm |
1.69 oz. | |
Screw thread ground joint, connecting cap, loosening ring | |
1 Case | |
To Provide Uniform Spray Mist of Visualization Reagents for TLC and Paper Chromatography | |
3 to 5 psi | |
6.85 in. | |
1.96 in. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Special Offers
-
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DWK Life Sciences Kimble™ Kontes™ Reagent Sprayers with Screw Thread Ground Joint > Capacity: 50mL; 19/22 standard taper joint
Spot an opportunity for improvement?Share a Content Correction