Learn More
DWK Life Sciences Kimble™ Score-Break Trimmed-Stem Ampules
Offer maximum solution stability, easy sealing and opening
£62.65
Product Code | Brand | Capacity (Metric) | Color | Diameter (English) Outer | Length (English) Body | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Color | Diameter (English) Outer | Length (English) Body | Price | Quantity & Availability | ||
17760858
|
DWK Life Sciences
12040G-2 |
2 mL | Amber | 0.47 in. | 1.18 in. |
£62.65
Pack of 144 |
Estimated Shipment: 21-02-2025
Log in to see stock. |
||
Description
- Choice of Amber 203 or Kimax-51™ clear
- United States Pharmacopeia (USP) Type I borosilicate glass
- Pull sealed
- Score-break, trimmed stem for easy opening
- Special ampule file not required
Amber ampules
- 1mL and 2mL
- 2mL and 20mL
Specifications
Borosilicate Glass | |
Prescored Ampule |