Learn More
DWK Life Sciences Kimble™ Screw Caps for Disposable Glass Tubes
Linerless, one-piece closures with inner sealing ring
Brand: DWK Life Sciences 73805B-13415
Code : A0
Additional Details : Weight : 1.00000kg
Description
Manufactured from polypropylene. Kimble Chase™ Screw Caps for Disposable Glass Tubes are autoclavable.
- Polypropylene
- Autoclavable
Compliance
Designed from ASTM™ E 982
Specifications
Screw Cap | |
ASTM E982 | |
Polypropylene | |
Disposable Glass Tubes |
13-415 | |
Autoclavable | |
Natural |