missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimberly-Clark™ Kimtech™ Opal™ Nitrile Gloves

Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
Description
- Kimtech™ Opal™ nitrile gloves guard against contamination by chemical splash and micro-organism hazards, delivering seamless protection when and where it counts.
- The powder-free gloves are ideal for use in research and production facilities, life sciences and non-sterile drug manufacturing applications.
- The gloves feature an innovative approach using a thin nitrile polymer with a tip thickness of just 0.07 mm, but with excellent tensile strength suitable for rigorous process use.
- Easy to don, soft and comfortable Kimtech™ Opal™ nitrile gloves are suitable for extended wearing time.
- The absence of natural rubber latex, sulphur and vulcanizing agents* reduces the risk of TYPE I and TYPE IV glove-associated skin reactions, protecting the wearer as well as the application.
- Precision manufacturing reduces the risk of contamination and ensures the nitrile gloves offer high levels of regulatory compliance.
- Beaded cuffs and textured fingertips enable excellent handling of both wet and dry materials, and the ambidextrous gloves have been extensively tested under stringent conditions.
- In addition, the thin nitrile construction and efficient packaging mean that the chemical-resistant gloves can reduce waste by up to 50%.
- Available in multiple sizes: X-Small (62880), Small (62881), Medium (62883), Large (62883), X-Large (62884)
*No sulphur added and formulated without the following vulcanising chemicals and accelerators: Thiurams, Thiazoles, Guanidines and Carbamates.
Greener Choice Claims
- Environmental benefits include:
- Recyclable
Specifications
Specifications
Certifications/Compliance | ISO 3 |
Certifications/Compliance (Europe) | PPE Cat III according to Regulation (EU) 2016/425, EN ISO 374-1 Type B, EN ISO 374-5 Micro Organism and Virus Protection |
Accelerator Free | Yes |
AQL | 1.5 |
Color | Dark Blue |
Disposable | Single-use |
Exterior Finish | Partially Textured |
Finger Thickness (English) | 1.8 mil |
Finger Thickness (Metric) | 0.18 mm |
Hand | Ambidextrous |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Documents
Product Information
Product Guides
CE Certificate
File size: 399.7 kB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Kimberly-Clark™ Kimtech™ Opal™ Nitrile Gloves >
Spot an opportunity for improvement?Share a Content Correction