missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimberly-Clark™ Kimtech™ Opal™ Nitrile Gloves
These powder free, ambidextrous Kimtech™ Opal™ Nitrile Gloves with beaded cuff and textured fingertips guard against contamination by chemical splash and micro-organism hazards while reducing the risk of TYPE I and IV glove-associated skin reactions.
£31.00 - £37.00
Specifications
Certifications/Compliance | ISO 3 |
---|---|
Certifications/Compliance (Europe) | PPE Cat III according to Regulation (EU) 2016/425, EN ISO 374-1 Type B, EN ISO 374-5 Micro Organism and Virus Protection |
Accelerator Free | Yes |
AQL | 1.5 |
Color | Dark Blue |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17367659
|
Kimberly-Clark™
62880 |
X-Small |
£37.00
Pack of 200 |
In Stock
Log in to see stock. |
|||||
17377659
|
Kimberly-Clark™
62881 |
Small |
£37.00
Pack of 200 |
In Stock
Log in to see stock. |
|||||
17387659
|
Kimberly-Clark™
62882 |
Medium |
£37.00
Pack of 200 |
In Stock
Log in to see stock. |
|||||
17397659
|
Kimberly-Clark™
62883 |
Large |
£37.00
Pack of 200 |
In Stock
Log in to see stock. |
|||||
17307669
|
Kimberly-Clark™
62884 |
X-Large |
£31.00
Pack of 170 |
In Stock
Log in to see stock. |
|||||
Description
- Kimtech™ Opal™ nitrile gloves guard against contamination by chemical splash and micro-organism hazards, delivering seamless protection when and where it counts.
- The powder-free gloves are ideal for use in research and production facilities, life sciences and non-sterile drug manufacturing applications.
- The gloves feature an innovative approach using a thin nitrile polymer with a tip thickness of just 0.07 mm, but with excellent tensile strength suitable for rigorous process use.
- Easy to don, soft and comfortable Kimtech™ Opal™ nitrile gloves are suitable for extended wearing time.
- The absence of natural rubber latex, sulphur and vulcanizing agents* reduces the risk of TYPE I and TYPE IV glove-associated skin reactions, protecting the wearer as well as the application.
- Precision manufacturing reduces the risk of contamination and ensures the nitrile gloves offer high levels of regulatory compliance.
- Beaded cuffs and textured fingertips enable excellent handling of both wet and dry materials, and the ambidextrous gloves have been extensively tested under stringent conditions.
- In addition, the thin nitrile construction and efficient packaging mean that the chemical-resistant gloves can reduce waste by up to 50%.
- Available in multiple sizes: X-Small (62880), Small (62881), Medium (62883), Large (62883), X-Large (62884)
*No sulphur added and formulated without the following vulcanising chemicals and accelerators: Thiurams, Thiazoles, Guanidines and Carbamates.
Specifications
ISO 3 | |
Yes | |
Dark Blue | |
Partially Textured | |
0.18 mm | |
Yes | |
2 mil | |
Powder Free | |
Non-sterile | |
0.05 mm | |
24 cm |
PPE Cat III according to Regulation (EU) 2016/425, EN ISO 374-1 Type B, EN ISO 374-5 Micro Organism and Virus Protection | |
1.5 | |
Single-use | |
1.8 mil | |
Ambidextrous | |
Nitrile | |
0.05 mm | |
Nitrile Examination Glove | |
2 mil | |
9.45 in. |